5-Cyano-2-methylpyridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
6-Methylnicotinonitrile, 5-Cyano-2-picoline
C3050-1G
C3050-5G
TCIAC3050-1GEA
33
EUR
TCIAC3050-1G
TCIAC3050-5G
5-Cyano-2-methylpyridine ≥98.0% (by GC, titration analysis)
5-Cyano-2-methylpyridine
Formula:
C₇H₆N₂ Peso molecolare: 118,14 g/mol Punto di ebollizione: 217°C |
MDL Number:
MFCD00038042 CAS Number: 3222-48-8 UN: 3439 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 82.0 to 86.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...