2-Chloro-4-methylphenylboronic acid (contains varying amounts of Anhydride)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Chloro-4-methylbenzeneboronic acid
C3049-1G
C3049-5G
TCIAC3049-1GEA
22
EUR
TCIAC3049-1G
TCIAC3049-5G
2-Chloro-4-methylphenylboronic acid (contains varying amounts of Anhydride)
2-Chloro-4-methylphenylboronic acid
Formula:
C₇H₈BClO₂ Peso molecolare: 170,40 g/mol Punto di fusione: 202°C |
MDL Number:
MFCD03411936 CAS Number: 145349-62-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 112.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...