3-Chloropropyl isocyanate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
C3027-1G
C3027-5G
TCIAC3027-1GEA
61
EUR
TCIAC3027-1G
TCIAC3027-5G
3-Chloropropyl isocyanate ≥98.0% (by GC)
3-Chloropropyl isocyanate
Formula:
C₄H₆ClNO Peso molecolare: 119,55 g/mol Punto di ebollizione: 154°C Flash Pt: 62°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00010384 CAS Number: 13010-19-0 UN: 2206 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...