3-Chloropyridine-2-carbonitrile ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
C3022-1G
C3022-5G
TCIAC3022-1GEA
23
EUR
TCIAC3022-1G
TCIAC3022-5G
3-Chloropyridine-2-carbonitrile ≥98.0% (by GC)
3-Chloropyridine-2-carbonitrile
Formula:
C₆H₃ClN₂ Peso molecolare: 138,56 g/mol Punto di ebollizione: 100°C |
MDL Number:
MFCD03788834 CAS Number: 38180-46-0 UN: 3439 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 81.0 to 85.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...