Stampa…

6-Chloro-2-pyridinecarboxaldehyde ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Chloropyridine-2-carboxaldehyde, 6-Chloro-2-formylpyridine, 6-Chloropicolinaldehyde

C3017-1G C3017-5G
TCIAC3017-1GEA 151 EUR
TCIAC3017-1G TCIAC3017-5G
6-Chloro-2-pyridinecarboxaldehyde ≥98.0% (by GC)
6-Chloro-2-pyridinecarboxaldehyde
Formula: C₆H₄ClNO
Peso molecolare: 141,55 g/mol
MDL Number: MFCD09832941
CAS Number: 54087-03-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 69.0 to 73.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR