Stampa…

4-Chloro-2,6-diphenylpyrimidine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

C3016-1G C3016-5G
TCIAC3016-1GEA 55 EUR
TCIAC3016-1G TCIAC3016-5G
4-Chloro-2,6-diphenylpyrimidine ≥98.0% (by GC)
4-Chloro-2,6-diphenylpyrimidine
Formula: C₁₆H₁₁ClN₂
Peso molecolare: 266,73 g/mol
MDL Number: MFCD00234890
CAS Number: 29509-91-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82c6eae21467d60626fabab65bf578291c40f2fb&vid=82c6eae21467d60626fabab65bf578291c40f2fb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 104.0 to 108.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR