4-Chloro-5'-phenyl-1,1':3',1''-terphenyl ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
5'-(4-Chlorophenyl)-1,1':3',1''-terphenyl
C3008-200MG
C3008-1G
TCIAC3008-200MGEA
41
EUR
TCIAC3008-200MG
TCIAC3008-1G
4-Chloro-5'-phenyl-1,1':3',1''-terphenyl ≥98.0% (by GC)
4-Chloro-5'-phenyl-1,1':3',1''-terphenyl
Formula:
C₂₄H₁₇Cl Peso molecolare: 340,85 g/mol |
CAS Number:
116941-51-6 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 141.0 to 145.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...