4-Chloro-D-mandelic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
(R)-2-(4-Chlorophenyl)-2-hydroxyacetic Acid
Formula:
C₈H₇ClO₃ Peso molecolare: 186.59 g/mol |
MDL Number:
MFCD00798430 CAS Number: 32189-36-9 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 119.0 to 123.0 deg-C |
Specific rotation [a]20/D | -132.0 to -140.0 deg (C=0.3,EtOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...