(S)-Benzyl-2-carbamoylpyrrolidine-1-carboxylate ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI
Formula:
C₁₃H₁₆N₂O₃ Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00020830 CAS Number: 34079-31-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 90.0 to 94.0 deg-C |
Specific rotation [a]20/D | -94.0 to -99.0 deg (C=0.5,CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...