Stampa…

2-Chlorobenzyl thiocyanate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

C3003-5G C3003-1G
TCIAC3003-5GEA 224 EUR
TCIAC3003-5G TCIAC3003-1G
2-Chlorobenzyl thiocyanate ≥98.0% (by GC)
2-Chlorobenzyl thiocyanate
Formula: C₈H₆ClNS
Punto di ebollizione: 140°C
MDL Number: MFCD09025671
CAS Number: 2082-66-8

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR