Stampa…

5-Chloro-2-formylpyridine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Chloropyridine-2-carbaldehyde, 5-Chloropicolinaldehyde, 5-Chloro-2-pyridinecarbaldehyde

C3000-1G C3000-5G
TCIAC3000-1GEA 38 EUR
TCIAC3000-1G TCIAC3000-5G
5-Chloro-2-formylpyridine ≥98.0% (by GC)
5-Chloro-2-formylpyridine
Formula: C₆H₄ClNO
Peso molecolare: 141,56 g/mol
MDL Number: MFCD08277271
CAS Number: 31181-89-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 63.0 to 67.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR