Stampa…

3-Chloroisonicotinonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

C2998-1G C2998-5G
TCIAC2998-1GEA 38 EUR
TCIAC2998-1G TCIAC2998-5G
3-Chloroisonicotinonitrile ≥98.0% (by GC)
3-Chloroisonicotinonitrile
Formula: C₆H₃ClN₂
Peso molecolare: 138,56 g/mol
MDL Number: MFCD05663706
CAS Number: 68325-15-5
UN: 3439
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 76.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR