Stampa…
5-Chloro-1H-pyrrolo[2,3-b]pyridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Attenzione
Synonyms: 5-Chloro-7-azaindole
C2996-1G C2996-5G
TCIAC2996-1GEA 57 EUR
TCIAC2996-1G TCIAC2996-5G
5-Chloro-1H-pyrrolo[2,3-b]pyridine ≥98.0% (by GC, titration analysis)
5-Chloro-1H-pyrrolo[2,3-b]pyridine
Formula: C₇H₅ClN₂
Peso molecolare: 152,58 g/mol
MDL Number: MFCD06659676
CAS Number: 866546-07-8
Visualizza i codici
Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 163.0 to 167.0 deg-C