Stampa…

3-Chloroisonicotinic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

C2983-5G C2983-25G
TCIAC2983-5GEA 70 EUR
TCIAC2983-5G TCIAC2983-25G
3-Chloroisonicotinic acid ≥98.0% (by GC, titration analysis)
3-Chloroisonicotinic acid
Formula: C₆H₄ClNO₂
MDL Number: MFCD03094685
CAS Number: 88912-27-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR