Stampa…

3-(Carbamimidoylthio)-1-propanesulphonic acid ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 3-(Carbamimidoylthio)propane-1-sulphonic acid, 3-(Amidinothio)-1-propanesulphonic acid

C2982-25G C2982-100G
TCIAC2982-25GEA 64 EUR
TCIAC2982-25G TCIAC2982-100G
3-(Carbamimidoylthio)-1-propanesulphonic acid ≥98.0% (by HPLC, total nitrogen)
3-(Carbamimidoylthio)-1-propanesulphonic acid
Formula: C₄H₁₀N₂O₃S₂
Peso molecolare: 198,26 g/mol
Punto di fusione: 257°C
MDL Number: MFCD00085942
CAS Number: 21668-81-5
EINECS: 244-520-8
UN: 3261
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR