Stampa…

5'-Chloro-2'-hydroxy-3'-nitroacetophenone ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-Chlor-3-ethanoyl-4-hydroxy-5-nitrobenzene, 1-(5-Chloro-2-hydroxy-3-nitrophenyl)ethanone

C2973-5G C2973-25G
TCIAC2973-5GEA 17 EUR
TCIAC2973-5G TCIAC2973-25G
5'-Chloro-2'-hydroxy-3'-nitroacetophenone ≥98.0% (by GC, titration analysis)
5'-Chloro-2'-hydroxy-3'-nitroacetophenone
Formula: C₈H₆ClNO₄
Peso molecolare: 215,59 g/mol
MDL Number: MFCD00191921
CAS Number: 84942-40-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 131.0 to 135.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR