4,4'-Carbonyldimorpholine ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₉H₁₆N₂O₃ |
MDL Number:
MFCD06200603 CAS Number: 38952-62-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 142.0 to 146.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...