Stampa…

3-Chlorophenethylamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 2-(3-Chlorophenyl)ethylamine

C2955-5G C2955-25G
TCIAC2955-5GEA 41 EUR
TCIAC2955-5G TCIAC2955-25G
3-Chlorophenethylamine ≥98.0% (by GC, titration analysis)
3-Chlorophenethylamine
Formula: C₈H₁₀ClN
Punto di ebollizione: 145°C
MDL Number: MFCD00047957
CAS Number: 13078-79-0

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR