Stampa…

1-(4-Chlorophenyl)cyclopropanecarboxylic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-(4-Chlorophenyl)-1-cyclopropanecarboxylic acid

C2954-5G C2954-25G
TCIAC2954-5GEA 58 EUR
TCIAC2954-5G TCIAC2954-25G
1-(4-Chlorophenyl)cyclopropanecarboxylic acid ≥98.0% (by titrimetric analysis)
1-(4-Chlorophenyl)cyclopropanecarboxylic acid
Formula: C₁₀H₉ClO₂
Peso molecolare: 196,64 g/mol
MDL Number: MFCD00001289
CAS Number: 72934-37-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 152.0 to 156.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR