Stampa…

4-Chloropicolinamide ≥97.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Chloropyridine-2-carboxamide

C2949-1G C2949-5G
TCIAC2949-1GEA 19 EUR
TCIAC2949-1G TCIAC2949-5G
4-Chloropicolinamide ≥97.0% (by HPLC, titration analysis)
4-Chloropicolinamide
Formula: C₆H₅ClN₂O
Peso molecolare: 156,57 g/mol
MDL Number: MFCD01085339
CAS Number: 99586-65-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 160.0 to 164.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR