Cefotetan ≥98.0% (by HPLC)
Fornitore: TCI
Formula:
C₁₇H₁₇N₇O₈S₄ Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00864983 CAS Number: 69712-56-7 EINECS: 274-093-3 Merck Index: 14,01934 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Specific rotation [a]20/D | +115.0 to +122.0 deg (C=1,0.5%NaHCO3 sol. (0.5%)) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...