Stampa…

7-Chloro-1,2,3,4-tetrahydrobenzo[b]azepin-5-one ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 7-Chloro-3,4-dihydro-1H-benzo[b]azepin-5(2H)-one, 7-Chloro-1,2,3,4-tetrahydro-5H-1-benzazepin-5-one

C2896-1G C2896-5G
TCIAC2896-1GEA 73 EUR
TCIAC2896-1G TCIAC2896-5G
7-Chloro-1,2,3,4-tetrahydrobenzo[b]azepin-5-one ≥98.0% (by GC, titration analysis)
7-Chloro-1,2,3,4-tetrahydrobenzo[b]azepin-5-one
Formula: C₁₀H₁₀ClNO
Peso molecolare: 195.6455 g/mol
MDL Number: MFCD06738694
CAS Number: 160129-45-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 103.0 to 107.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR