Stampa…

2'-Cyanoacetanilide ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Cyanoacetanilide, 2-Cyano-N-phenylacetamide

C2894-1G C2894-5G
TCIAC2894-1GEA 28 EUR
TCIAC2894-1G TCIAC2894-5G
2'-Cyanoacetanilide ≥98.0% (by GC)
2'-Cyanoacetanilide
Formula: C₉H₈N₂O
Peso molecolare: 160,1726 g/mol
MDL Number: MFCD00019840
CAS Number: 621-03-4
EINECS: 210-666-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 198.0 to 202.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR