Stampa…

1-(3-Chloroanilino)-4-phenylphthalazine ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: N-(3-Chlorophenyl)-4-phenylphthalazin-1-amine, MY-5445, 1-(3-Chlorophenyl)amino-4-phenylphthalazine

C2884-20MG C2884-100MG
TCIAC2884-20MGEA 57 EUR
TCIAC2884-20MG TCIAC2884-100MG
1-(3-Chloroanilino)-4-phenylphthalazine ≥98.0% (by GC)
1-(3-Chloroanilino)-4-phenylphthalazine
Formula: C₂₀H₁₄ClN₃
Peso molecolare: 331,80 g/mol
MDL Number: MFCD00209838
CAS Number: 78351-75-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR