3-Chloro-4-(3-fluorobenzyloxy)nitrobenzene ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
2-Chloro-1-((3-fluorobenzyl)oxy)-4-nitrobenzene, 2-Chloro-1-(3-fluorobenzyloxy)-4-nitrobenzene
C2875-1G
C2875-5G
TCIAC2875-1GEA
28
EUR
TCIAC2875-1G
TCIAC2875-5G
3-Chloro-4-(3-fluorobenzyloxy)nitrobenzene ≥98.0% (by GC)
3-Chloro-4-(3-fluorobenzyloxy)nitrobenzene
Formula:
C₁₃H₉ClFNO₃ Peso molecolare: 281,67 g/mol |
MDL Number:
MFCD06809827 CAS Number: 443882-99-3 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 99.0 to 103.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...