3-Chloro-4-((3-fluorobenzyl)oxy)aniline ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-Chloro-4-(3-fluorobenzyloxy)aniline
C2874-1G
C2874-5G
TCIAC2874-1GEA
31
EUR
TCIAC2874-1G
TCIAC2874-5G
3-Chloro-4-((3-fluorobenzyl)oxy)aniline ≥98.0% (by GC, titration analysis)
3-Chloro-4-((3-fluorobenzyl)oxy)aniline
Formula:
C₁₃H₁₁ClFNO Peso molecolare: 251,69 g/mol |
MDL Number:
MFCD06809822 CAS Number: 202197-26-0 EINECS: 445-590-4 UN: 3077 ADR: 9,III |
Risultati del test di specificazione
Appearance | White to Gray to Red powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 78.0 to 82.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...