Stampa…

6-Chloro-4-hydroxycoumarin ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Chloro-4-hydroxy-2H-chromen-2-one

C2870-1G C2870-5G
TCIAC2870-1GEA 27 EUR
TCIAC2870-1G TCIAC2870-5G
6-Chloro-4-hydroxycoumarin ≥97.0% (by GC, titration analysis)
6-Chloro-4-hydroxycoumarin
Formula: C₉H₅ClO₃
Peso molecolare: 196,59 g/mol
MDL Number: MFCD00239399
CAS Number: 19484-57-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystaline
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR