6-Chloro-3-formylchromone ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
6-Chloro-4-oxo-4H-chromene-3-carbaldehyde
C2869-5G
C2869-25G
TCIAC2869-5GEA
36
EUR
TCIAC2869-5G
TCIAC2869-25G
6-Chloro-3-formylchromone ≥98.0% (by GC)
6-Chloro-3-formylchromone
Formula:
C₁₀H₅ClO₃ Peso molecolare: 208,597 g/mol |
MDL Number:
MFCD00139138 CAS Number: 42248-31-7 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 96.0 % |
Melting point | 165.0 to 169.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...