Stampa…

Clotrimazole ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-(2-Chlorotrityl)imidazole, 1-[(2-Chlorophenyl)diphenylmethyl]imidazole

C2867-5G C2867-25G
TCIAC2867-5GEA 28 EUR
TCIAC2867-5G TCIAC2867-25G
Clotrimazole ≥98.0% (by HPLC, titration analysis)
Clotrimazole
Formula: C₂₂H₁₇ClN₂
Peso molecolare: 344,84 g/mol
MDL Number: MFCD00057220
CAS Number: 23593-75-1
EINECS: 245-764-8
Merck Index: 14,02417

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 144.0 to 148.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR