Stampa…

6-Chloro-1,2,3,4-tetrahydrocarbazole ≥98.0% (by GC)

Fornitore: TCI
Attenzione

C2857-1G C2857-5G
TCIAC2857-1GEA 47 EUR
TCIAC2857-1G TCIAC2857-5G
6-Chloro-1,2,3,4-tetrahydrocarbazole ≥98.0% (by GC)
6-Chloro-1,2,3,4-tetrahydrocarbazole
Formula: C₁₂H₁₂ClN
MDL Number: MFCD00541331
CAS Number: 36684-65-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 147.0 to 151.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR