Stampa…

Cedrol ≥98.0% (by GC)

Fornitore: TCI
Attenzione

C2854-5G C2854-25G
TCIAC2854-5GEA 60 EUR
TCIAC2854-5G TCIAC2854-25G
Cedrol ≥98.0% (by GC)
Cedrol
Formula: C₁₅H₂₆O
Punto di ebollizione: 273°C
MDL Number: MFCD00062952
CAS Number: 77-53-2
EINECS: 201-035-6
Merck Index: 14,01911

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 84.0 to 88.0 deg-C
Specific rotation [a]20/D +9.0 to +11.0 deg (C=5, CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR