2-(((Benzyloxy)carbonyl)(methyl)amino)acetic acid ≥97.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
N-Cbz-N-methylglycine, Z-N-Me-Gly-OH, Z-Sar-OH, N-((Benzyloxy)carbonyl)-N-methylglycine, N-(Benzyloxycarbonyl)sarcosine
C2848-25G
C2848-5G
TCIAC2848-25GEA
65
EUR
TCIAC2848-25G
TCIAC2848-5G
2-(((Benzyloxy)carbonyl)(methyl)amino)acetic acid ≥97.0% (by HPLC)
2-(((Benzyloxy)carbonyl)(methyl)amino)acetic acid
Formula:
C₁₁H₁₃NO₄ Peso molecolare: 223,3 g/mol |
MDL Number:
MFCD00021747 CAS Number: 39608-31-6 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid to cloudy liquid |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 90.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...