7-Chloroisatin ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
7-Chloroindoline-2,3-dione, 7-Chloro-1H-indole-2,3-dione, 7-Cloroindoline-2,3-dione
C2840-1G
C2840-5G
TCIAC2840-1GEA
23
EUR
TCIAC2840-1G
TCIAC2840-5G
7-Chloroisatin ≥98.0% (by GC, titration analysis)
7-Chloroisatin
Formula:
C₈H₄ClNO₂ Peso molecolare: 181,58 g/mol |
MDL Number:
MFCD00022796 CAS Number: 7477-63-6 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 187.0 to 191.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...