Stampa…

(S)-6-Chloro-4-hydroxy-3,4-dihydro-2H-thieno[3,2-e][1,2]thiazine-1,1-dioxide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: (S)-6-Chloro-4-hydroxy-3,4-dihydro-2H-thieno[3,2-e][1,2]thiazine 1,1-dioxide

C2832-1G C2832-5G
TCIAC2832-1GEA 111 EUR
TCIAC2832-1G TCIAC2832-5G
(S)-6-Chloro-4-hydroxy-3,4-dihydro-2H-thieno[3,2-e][1,2]thiazine-1,1-dioxide ≥98.0% (by HPLC, titration analysis)
(S)-6-Chloro-4-hydroxy-3,4-dihydro-2H-thieno[3,2-e][1,2]thiazine-1,1-dioxide
Formula: C₆H₆ClNO₃S₂
Peso molecolare: 239,69 g/mol
MDL Number: MFCD09033311
CAS Number: 160982-16-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 128.0 to 132.0 deg-C
Specific rotation [a]20/D -4.0 to -6.0 deg (C=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR