Stampa…

3-Coumaranone ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Benzofuran-3(2H)-one

C2825-1G C2825-5G
TCIAC2825-1GEA 27 EUR
TCIAC2825-1G TCIAC2825-5G
3-Coumaranone ≥98.0% (by GC)
3-Coumaranone
Formula: C₈H₆O₂
Peso molecolare: 134,13 g/mol
MDL Number: MFCD00051810
CAS Number: 7169-34-8

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 98.0 to 102.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR