Stampa…

ɑ-Cyclohexylphenylacetic acid ≥95.0% (by GC, titration analysis)

Fornitore: TCI

Synonyms: ['2-Cyclohexyl-2-phenylacetic acid'], α-Cyclohexylphenylacetic acid, 2-Cyclohexyl-2-phenylacetic Acid

C2812-1G C2812-5G
TCIAC2812-1GEA 26 EUR
TCIAC2812-1G TCIAC2812-5G
ɑ-Cyclohexylphenylacetic acid ≥95.0% (by GC, titration analysis)
alpha-Cyclohexylphenylacetic acid
Formula: C₁₄H₁₈O₂
Peso molecolare: 218.30 g/mol
MDL Number: MFCD00001477
CAS Number: 3894-09-5
EINECS: 223-443-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 150.0 to 154.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR