Stampa…

4-Chloroisatoic anhydride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 7-Chloro-1H-benzo[d][1,3]oxazine-2,4-dione, 7-Chloro-2H-3,1-benzoxazine-2,4(1H)-dione

C2790-1G C2790-5G
TCIAC2790-1GEA 34 EUR
TCIAC2790-1G TCIAC2790-5G
4-Chloroisatoic anhydride ≥98.0% (by HPLC, titration analysis)
4-Chloroisatoic anhydride
Formula: C₈H₄ClNO₃
Peso molecolare: 197,58 g/mol
MDL Number: MFCD00090431
CAS Number: 40928-13-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR