Stampa…

CZBDF ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: 3,7-Bis[4-(9H-carbazol-9-yl)phenyl]-2,6-diphenylbenzo[1,2-b:4,5-b']difuran, 9,9'-[(2,6-Diphenylbenzo[1,2-b:4,5-b']difuran-3,7-diyl)bis(4,1-phenylene)]bis(9H-carbazole)

C2780-200MG C2780-1G
TCIAC2780-200MGEA 97 EUR
TCIAC2780-200MG TCIAC2780-1G
CZBDF ≥98.0% (by HPLC)
CZBDF
Formula: C₅₈H₃₆N₂O₂
Peso molecolare: 792,94 g/mol
MDL Number: MFCD28100824
CAS Number: 1092578-51-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR