Stampa…

1-Cyclopropyl-6,7-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic acid

C2779-1G C2779-5G
TCIAC2779-1GEA 67 EUR
TCIAC2779-1G TCIAC2779-5G
1-Cyclopropyl-6,7-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
1-Cyclopropyl-6,7-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Formula: C₁₃H₉F₂NO₃
Peso molecolare: 265.2123 g/mol
Punto di fusione: 289°C
MDL Number: MFCD01646375
CAS Number: 93107-30-3
EINECS: 413-760-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR