6-Chloro-o-toluic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Chloro-6-methylbenzoic acid
C2776-1G
C2776-5G
TCIAC2776-1GEA
56
EUR
TCIAC2776-1G
TCIAC2776-5G
6-Chloro-o-toluic acid ≥98.0% (by GC, titration analysis)
6-Chloro-o-toluic acid
Formula:
C₈H₇ClO₂ Peso molecolare: 170,6 g/mol |
MDL Number:
MFCD00045799 CAS Number: 21327-86-6 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light red powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 104.0 to 108.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...