4-Chloro-6,7-bis(2-methoxyethoxy)quinazoline ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI
|
Attenzione
|
C2774-5G
C2774-25G
TCIAC2774-5GEA
50
EUR
TCIAC2774-5G
TCIAC2774-25G
4-Chloro-6,7-bis(2-methoxyethoxy)quinazoline ≥98.0% (by HPLC, total nitrogen)
4-Chloro-6,7-bis(2-methoxyethoxy)quinazoline
Formula:
C₁₄H₁₇ClN₂O₄ Peso molecolare: 312.7500 g/mol |
MDL Number:
MFCD09751265 CAS Number: 183322-18-1 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 99.0 to 104.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...