Stampa…

2-(1-Methylguanidino)ethyl dihydrogen phosphate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Creatinol phosphate, Aplodan, 2-(1-Methylguanidino)ethyl dihydrogen phosphate

C2767-5G C2767-25G
TCIAC2767-5GEA 27 EUR
TCIAC2767-5G TCIAC2767-25G
2-(1-Methylguanidino)ethyl dihydrogen phosphate ≥98.0% (by HPLC, titration analysis)
2-(1-Methylguanidino)ethyl dihydrogen phosphate
Formula: C₄H₁₂N₃O₄P
Peso molecolare: 197,1295 g/mol
Punto di ebollizione: 244°C
MDL Number: MFCD00868297
CAS Number: 6903-79-3
EINECS: 230-011-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR