Stampa…

N-Carbobenzoxy-6-aminohexanoic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N-Cbz-epsilon-aminocaproic Acid, N-Cbz-6-aminohexanoic Acid, N-Benzyloxycarbonyl-6-aminohexanoic acid, N-Carbobenzoxy-epsilon-aminocaproic Acid, Z-ε-Acp-OH

C2757-5G C2757-25G
TCIAC2757-5GEA 39 EUR
TCIAC2757-5G TCIAC2757-25G
N-Carbobenzoxy-6-aminohexanoic acid ≥98.0% (by HPLC, titration analysis)
N-Carbobenzoxy-6-aminohexanoic acid
Formula: C₁₄H₁₉NO₄
Peso molecolare: 265,31 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00004423
CAS Number: 1947-00-8
EINECS: 217-748-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 56.0 to 60.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR