(-)-Corey lactone-4-phenylbenzoate ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
(3aR,4S,5R,6aS)-4-(Hydroxymethyl)-2-oxohexahydro-2H-cyclopenta[b]furan-5-yl 4-Phenylbenzoate, (3aR,4S,5R,6aS)-Hexahydro-4-(hydroxymethyl)-5-(4-phenylbenzoyloxy)-2H-cyclopenta[b]furan-2-one, (3aR,4S,5R,6aS)-4-(Hydroxymethyl)-2-oxohexahydro-2H-cyclopenta[b]furan-5-yl Biphenyl-4-carboxylate
Formula:
C₂₁H₂₀O₅ Peso molecolare: 352,39 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00078077 CAS Number: 31752-99-5 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Carbon) | 69.00 to 74.00 % |
Melting point | 131.0 to 135.0 deg-C |
Specific rotation [a]20/D | -81.0 to -85.0 deg (C=1,CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...