2-Chloro-5-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
C2755-200MG
C2755-1G
TCIAC2755-200MGEA
49
EUR
TCIAC2755-200MG
TCIAC2755-1G
2-Chloro-5-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
2-Chloro-5-methoxybenzoic acid
Formula:
C₈H₇ClO₃ |
MDL Number:
MFCD00466261 CAS Number: 6280-89-3 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 172.0 to 176.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...