Stampa…

Cetilistat ≥98.0% (by HPLC)

Fornitore: TCI

C2745-1G C2745-5G
TCIAC2745-1GEA 37 EUR
TCIAC2745-1G TCIAC2745-5G
Cetilistat ≥98.0% (by HPLC)
Cetilistat
Formula: C₂₅H₃₉NO₃
MDL Number: MFCD09839697
CAS Number: 282526-98-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Melting point 72.0 to 76.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR