Stampa…

5-Chloroisatoic anhydride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 6-Chloro-1H-benzo[d][1,3]oxazine-2,4-dione

C2743-5G C2743-25G
TCIAC2743-5GEA 50 EUR
TCIAC2743-5G TCIAC2743-25G
5-Chloroisatoic anhydride ≥98.0% (by HPLC, titration analysis)
5-Chloroisatoic anhydride
Formula: C₈H₄ClNO₃
MDL Number: MFCD00006701
CAS Number: 4743-17-3
EINECS: 225-255-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR