2-Chloro-4-(methylsulphonyl)benzoic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-Chloro-4-methylsulfonylbenzoic acid, 2-Chloro-4-methylsulphonylbenzoic acid
C2726-5G
C2726-25G
TCIAC2726-5GEA
24
EUR
TCIAC2726-5G
TCIAC2726-25G
2-Chloro-4-(methylsulphonyl)benzoic acid ≥98.0% (by GC, titration analysis)
2-Chloro-4-(methylsulphonyl)benzoic acid
Formula:
C₈H₇ClO₄S Peso molecolare: 234,66 g/mol |
MDL Number:
MFCD00216496 CAS Number: 53250-83-2 EINECS: 406-520-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 194.0 to 198.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...