Stampa…

2-Chloro-1,4-naphthoquinone ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Chloronaphthalene-1,4-dione

C2721-5G C2721-25G
TCIAC2721-5GEA 95 EUR
TCIAC2721-5G TCIAC2721-25G
2-Chloro-1,4-naphthoquinone ≥98.0% (by GC)
2-Chloro-1,4-naphthoquinone
Formula: C₁₀H₅ClO₂
Peso molecolare: 192,60 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00029187
CAS Number: 1010-60-2
EINECS: 213-776-2

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 114.0 to 118.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR