4-Chlorobutyraldehyde diethyl acetal ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-Chloro-1,1-diethoxybutane, 4-Chlorobutanal diethyl acetal
C2717-5G
C2717-25G
TCIAC2717-5GEA
28
EUR
TCIAC2717-5G
TCIAC2717-25G
4-Chlorobutyraldehyde diethyl acetal ≥97.0% (by GC)
4-Chlorobutyraldehyde diethyl acetal
Formula:
C₈H₁₇ClO₂ Peso molecolare: 180,67 g/mol Punto di ebollizione: 92°C |
MDL Number:
MFCD00671476 CAS Number: 6139-83-9 UN: 1993 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Orange to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...